Learn More
Description
- Form: Off-white solid
- Molar Mass: 443.4
- Purity: Homogeneous single spot by TLC
- Soluble in dH2O or DMSO
- Synonyms: Sulfobiotin-NHS, Sulfosuccinimidobiotin
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | Biotin-NHS, Water-Soluble |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.