Learn More
Description
- Molar Mass: 341.4
- Purity: ≥90% by NMR
- Synonyms: Biotin N-hydroxysuccinimide Ester
Specifications
Specifications
| Quantity | 250 mg |
| Product Type | Biotin-NHS conjugate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.