Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Biotin-X-NHS, Water-Soluble

Catalog No. 20318950MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-318-950MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-318-950MG Supplier MilliporeSigma™ Supplier No. 20318950MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For labeling antibodies, hormones or other polypeptide molecules containing lysine residues

  • Form: Off white to tan solid
  • Molar Mass: 556.6
  • Purity: ≥94% by HPLC
  • Soluble in dH2O or DMSO
  • Synonyms: Sulfobiotin-X-NHS, Sulfosuccinimidyl-6-(biotinamido)hexanoate
  • Ideal for labeling antibodies, hormones, or other polypeptide molecules containing lysine residues
  • Extended spacer arm provides maximum accessibility of biotin for avidin conjugates
TRUSTED_SUSTAINABILITY

Specifications

Description Biotin-X-NHS, Water-Soluble
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.