Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ DNP-Hemocyanin Conjugate, Keyhole Limpet

Catalog No. 324121100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-412-1100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-412-1100MG Supplier MilliporeSigma™ Supplier No. 324121100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For production of antibodies specific for DNP and hemocyanin

  • Contains ≥400 DNP-groups per molecule of protein
  • Form: Yellow fluffy solid
  • Soluble in water
TRUSTED_SUSTAINABILITY

Specifications

Product Type DNP-Hemocyanin Conjugate, Keyhole Limpet
Quantity 100 mg
Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.