Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Freund's Complete Adjuvant, Modified, Mycobacterium butyricum

Catalog No. 3442891SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-428-91SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-428-91SET Supplier MilliporeSigma™ Supplier No. 3442891SET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clear amber to slightly cloudy bacterial suspension

  • Composition: Each vial contains 10mL of Freund's complete adjuvant homogenized in following proportions by weight: 85% Drakeol 5NF, 15% Arlacel A (mannide monooleate emulsifier) and 0.1% Mycobacterium butyricum dry cells
  • Formulation: Yellow, oily, bacterial suspension
  • Quantity: 10 vials/set
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Set
Content And Storage 2°C to 8°C
Product Type Freund's Complete Adjuvant, Modified, Mycobacterium butyricum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.