Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Glutathione Assay Kit

Catalog No. 354102100T Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-410-2100T 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 35-410-2100T Supplier MilliporeSigma™ Supplier No. 354102100T
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specific for reduced glutathione (GSH) or total mercaptans

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Glutathione Assay Kit
Detection Method Colorimetric
Includes A Chromogen, 30% NaOH, Buffer, and a user protocol.
Quantity 100 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.