Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Glutathione Peroxidase Assay Kit

Catalog No. 3539191KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-391-91KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 35-391-91KIT Supplier MilliporeSigma™ Supplier No. 3539191KIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Measures glutathione peroxidase (GPx) activity in plasma, erythrocyte lysates, tissue homogenates and cell lysates

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Description Glutathione Peroxidase Assay Kit
Detection Method Colorimetric
For Use With (Application) Measures glutathione peroxidase (GPx) activity in plasma, erythrocyte lysates, tissue homogenates and cell lysates
Includes Assay Buffer, Sample Buffer, Glutathione Peroxidase (Control), Co-Substrate Mixture, Cumene Hydroperoxide, 96 Well Plate, Plate Cover, and a user protocol.
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.