Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Human Recombinant P-Selectin

Catalog No. 56130650UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-130-650UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-130-650UG Supplier MilliporeSigma™ Supplier No. 56130650UG
Add to Cart
Add to Cart

Recombinant, human P-selectin consisting of 730aa minus the transmembrane and cytoplasmic domains expressed in CHO cells

TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from Dulbecco's D-PBS (containing calcium and magnesium), 1% sorbitol
Quantity 50 μg
Source CHO cells
Storage Requirements -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.