Learn More
Description
PANSORBIN cells are useful as a solid-phase IgG-binding reagent due to the high-affinity interaction of protein A with the Fc domain of IgG. They work better when the antibody is human (IgG1, IgG2, IgG4), rabbit IgG (all isotypes), or mouse (IgG2a, IgG2b, IgG3).
- Activity: ≥2.0 mg human IgG/100mg cells
- Form: Homogeneous, milky suspension
- Formulation: Supplied as a 10% suspension of Staphylococcus aureus cells in PBS, 0.1% NaN3, pH 7.2
- Contaminants: Viable cells: ≤2500 cells/mL
- Storage: Store at 2° to 8°C. Do not freeze
Specifications
Specifications
| Product Type | PANSORBIN Cells |
| Content And Storage | 2°C to 8°C |
| Quantity | 10 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.