Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protease Assay Kit

Catalog No. 5391251KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit (200 Tests)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-912-51KIT 1 Kit (200 Tests)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-912-51KIT Supplier MilliporeSigma™ Supplier No. 5391251KIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For quantitative array of wide variety of proteases in biological samples, utilizing FTC-casein as substrate

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage FTC-casein should be stored at -20°C and is stable for one year from the date of purchase, Protease positive control should be stored at -20°C, Incubation buffer and assay buffer may be stored at 4deg.C or -20°C
Description Protease Assay Kit
Detection Method Colorimetric
For Use With (Application) For quantitative array of wide variety of proteases in biological samples, utilizing FTC-casein as substrate
Includes A Fluorescent-Casein Substrate, Incubation Buffer, Assay Buffer Trypsin Standard and a user protocol.
Quantity 1 Kit (200 Tests)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.