Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protein A, Peroxidase Conjugate

Catalog No. 5392531MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-925-31MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-925-31MG Supplier MilliporeSigma™ Supplier No. 5392531MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Native protein A conjugated to horseradish peroxidase

  • Form: Liquid
  • Formulation: In 10mM PBS, 0.5mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6
  • Purity: Single band 50,000 on SDS-PAGE
  • Preservative: 0.02% thimerosal
  • Applications: ELISA, Immunoblotting (Western Blotting)
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein A, Peroxidase Conjugate
Quantity 1 mg
Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.