Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protein G, Alkaline Phosphatase Conjugate

Catalog No. 539305500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-930-5500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-930-5500UG Supplier MilliporeSigma™ Supplier No. 539305500UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protein G conjugated to alkaline phosphatase

  • Does not react with IgA, IgE, or IgM
  • Form: Liquid
  • Formulation: In 50mM Tris buffer, 1mM MgCl2, 0.1mM ZnCl2, 1% BSA, 0.1% sodium azide, pH 7.6
  • Purity: Single band at 33.8 kd by SDS-PAGE
  • Preservative: ≤0.1% sodium azide
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein G, Alkaline Phosphatase Conjugate
Quantity 500 μg
Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.