Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Recombinant Human Furin

Catalog No. 34493250UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-493-250UL 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-493-250UL Supplier MilliporeSigma™ Supplier No. 34493250UL
Add to Cart
Add to Cart

xpressed in vaccinia virus (VV:hFUR713t) infected BSC-40 cells

Recombinant, human furin expressed in vaccinia virus (VV:hFUR713t) infected BSC-40 cells. Supplied as a crude preparation of soluble furin obtained from cytoplasmic extracts. Furin is a type I membrane protein that processes stromelysin-3 (MMP-11) and MT1-MMP. It cleaves pro-hormones, neuropeptide precursors, some viral proteins, and bacterial toxins.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 20 mM Bis-Tris buffer, 1mg/ml BSA, pH 7.0
Quantity 50 μL
Source Vaccinia virus
Storage Requirements ≤-70°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.