Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Streptavidin

Catalog No. 1897301MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-973-01MG 1 mg
18-973-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-973-01MG Supplier MilliporeSigma™ Supplier No. 1897301MG
Add to Cart
Add to Cart

High-affinity biotin binding protein

  • Form: Lyophilized solid
  • Molar Mass; 60,000
  • Specific Activity: ≥10 units/mg protein
  • Synonyms: Avidin, Streptomyces avidinii
  • Soluble in water or many low ionic strength buffers at neutral pH
  • Storage: Store at -20°C and protect from moisture
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Content And Storage -20°C
Product Type Streptavidin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.