Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Streptavidin, β-Galactosidase Conjugate

Catalog No. 5694042ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-940-42ML 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-940-42ML Supplier MilliporeSigma™ Supplier No. 5694042ML
Add to Cart
Add to Cart

For Enzyme immunoassay and immunohistochemistry

  • Form: Liquid
  • Formulation: In 100mM glycine, 100mM MgCl2, 50mM PBS, 1% BSA, pH 6.0
  • Preservative: ≤0.1% sodium azide
TRUSTED_SUSTAINABILITY

Specifications

Description Streptavidin, β-Galactosidase Conjugate
Quantity 2 mL
Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.