Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Trypsin Bovine MS Grade

Catalog No. 6502771SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 x 20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-027-71SET 5 x 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-027-71SET Supplier MilliporeSigma™ Supplier No. 6502771SET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A chemically modified trypsin that is resistant to autolysis and degradation

Trypsin specifically hydrolyzes amides, peptides, and proteins at the C-terminus side of arginine and lysine residues. This mass spectrometry grade trypsin is methylated, TPCK treated, and affinity purified. It is specifically designed for digestion of proteins for sequencing and peptide fragment analysis by mass spectrometry. A trypsin to protein ratio of 1:100 to 1:20 is recommended for mass spectrometry protocols. Note: 1 SET = 5 X 20μg of modified trypsin and 1mL of resuspension buffer.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
For Use With (Application) Digestion of proteins for sequencing and peptide fragment analysis by mass spectrometry
Quantity 5 x 20 μg
Source Bovine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.