Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calmodulin, Biotinylated, Porcine Brain, Protein

Catalog No. 208780100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-878-0100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-878-0100UG Supplier MilliporeSigma™ Supplier No. 208780100UG
Calmodulin (CaM) purified from porcine brain and biotinylated using NHS ester-activated biotin with an 8 atom spacer arm. A ubiquitous, calcium-binding protein that interacts with and regulates a wide variety of proteins, thus affecting different cellular functions and calcium-calmodulin-dependent signaling pathways. CaM mediates several biological processes, such as inflammation, apoptosis, muscle contraction, intracellular movement, short-term and long-term memory, nerve growth, and the immune response. Useful for enzyme activation, CaM binding studies, and general assessment of CaM-dependent signaling pathways. Biotin/Protein Ratio: 0.5-1.0 as determined by HABA assay.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 10mM sodium phosphate buffer, 5% sucrose, pH 7.0.
Preparation Method H2O (1mg/mL)
Purification Method SDS-PAGE
Quantity 100 μg
Immunogen Calmodulin
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 6 months at −20°C.
Regulatory Status RUO
Conjugate Biotin
Species Porcine
Recombinant Native
Form Lyophilized
Purity or Quality Grade >98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.