Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calmodulin, HisTag™ Human, Recombinant Protein

Catalog No. 20867050UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-867-050UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-867-050UG Supplier MilliporeSigma™ Supplier No. 20867050UG
Add to Cart
Add to Cart
Full-length, recombinant human calmodulin fused to a His·Tag™ sequence at the N-terminus. Contains four functional Ca2+-binding sites (aa 20-31; aa 56-67; aa 93-104; aa 129-140) with EF-hands (aa 7-42; aa 43-78; aa 80-115; aa 116-148); and a ubiquitination site at Lys21. This preparation is qualified for use as a substrate for protein tyrosine kinases in in vitro assays. Through its interaction with Ca2+, calmodulin mediates the control of a large number of enzymes, including protein kinases and phosphatases. It is expected to be phosphorylated at Thr44 by CaMK4. In addition, it serves as a substrate for various protein tyrosine kinases.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In PBS, 0.2% Protease Inhibitor Cocktail Set VII.
Purification Method SDS-PAGE
Quantity 50 μg
Immunogen Calmodulin
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Conjugate His-Tag
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.