Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calpain-1, Porcine Erythrocytes, Protein

Catalog No. 2087121MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-871-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-871-21MG Supplier MilliporeSigma™ Supplier No. 2087121MG
Add to Cart
Add to Cart
Native calpain-1 from porcine erythrocytes. Ca2+-dependent heterodimeric cysteine proteinase with low Ca2+ requirement (EC50= 2μM). Participates in the ATP release reaction of platelets stimulated with thrombin.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 20mM imidazole-HCl, 5mM β-mercaptoethanol, 1mM EDTA, 1mM EGTA, 30% glycerol, pH 6.8.
Quantity 1 mg
Immunogen Calpain-1
Storage Requirements Following initial thaw, aliquot and freeze (−70°C). Short-term storage of aliquots at 4°C or on ice is not recommended.
Regulatory Status RUO
Species Porcine
Recombinant Native
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.