Learn More
MilliporeSigma™ Casein Kinase II, Human, Recombinant Protein, E. coli
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | In 350mM NaCl, 20mM Tris-HCl, 2mM DTT, 1mM EDTA, 0.1% TRITON™ X-100 Detergent, pH 7.5. |
| Quantity | 10kU |
| Source | E. coli |
| Immunogen | Casein Kinase II |
| Storage Requirements | Following initial thaw, aliquot and freeze (−70°C). |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Recombinant |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.