Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Casein Kinase II, α-Subunit, Human, Recombinant Protein, E. coli

Catalog No. 21870410MIU Shop All MilliporeSigma Products
Change view
Click to view available options
10mIU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
21-870-410MIU 10mIU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-870-410MIU Supplier MilliporeSigma™ Supplier No. 21870410MIU
Recombinant, human casein kinase II α-subunit expressed in E. coli. The catalytic subunit of serine/threonine protein kinase CKII. Suitable for use in reactions where autophosphorylation of the regulatory CKII β-subunit is not desired. Shown to be extremely salt-sensitive with respect to its activity.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 500mM NaCl, 25mM Tris-HCl, 1mM DTT, 500μM PMSF, 50% glycerol, pH 8.5.
Purification Method SDS-PAGE
Quantity 10mIU
Source E. coli
Immunogen Casein Kinase II, α-Subunit
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.