Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Caspase-1, Human, Recombinant Protein, E. coli

Catalog No. 2187833000U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
3000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-878-33000U 3000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-878-33000U Supplier MilliporeSigma™ Supplier No. 2187833000U
Add to Cart
Add to Cart
Recombinant, human caspase-1 fused at the N-terminus to a His·Tag™ sequence and expressed in E. coli. Useful for the study of enzyme regulation, cleavage of target substrates, and inhibitor screening. M.W. 10000 and 20000.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 100mM NaCl, 50mM HEPES, 10mM DTT, 1mM EDTA, 10% glycerol, 0.5% CHAPS, pH 7.4.
Purification Method SDS-PAGE
Quantity 3000 U
Source E. coli
Immunogen Caspase-1
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.