Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cathepsin D, Human Liver, Protein

Catalog No. 21940115U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
15 U
30 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-940-115U 15 U
21-940-130U 30 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-940-115U Supplier MilliporeSigma™ Supplier No. 21940115U
Add to Cart
Add to Cart
Native cathepsin D from human liver. A major lysosomal aspartyl protease produced as a 52kDa proenzyme. Overexpression of cathepsin D in human breast cancers is associated with higher risk of relapse and metastasis. Cathepsin D degrades extracellular matrix and releases growth factors bound to the matrix.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 2mM sodium phosphate buffer, pH 6.5.
Preparation Method Sterile H2O or aqueous buffers. Please see vial label for lot-specific reconstitution volume.
Purification Method SDS-PAGE
Quantity 15 U
Immunogen Cathepsin D
Storage Requirements Following reconstitution, aliquot and freeze (−20°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 1 week at 4°C. Keep on ice when working with material. When product is brought to its active pH (3.3), autolysis occurs.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.