Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cathepsin L, Human Liver, Protein

Catalog No. 21940225UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-940-225UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-940-225UG Supplier MilliporeSigma™ Supplier No. 21940225UG
Add to Cart
Add to Cart
Native cathepsin L from human liver. The most potent of the lysosomal proteinases, having a higher activity than cathepsins B and H in the degradation of a variety of physiological protein substrates. Cathepsin L is believed to be responsible for the generation of endostatin from NC1 domain in collagen XVII.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 400mM NaCl, 20mM malonate buffer, 1mM EDTA, pH 5.5.
Quantity 25 μg
Immunogen Cathepsin L
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.