Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cathepsin V, HisTag™, Human, Recombinant Protein, NSO Cells

Catalog No. 21946710UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-946-710UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-946-710UG Supplier MilliporeSigma™ Supplier No. 21946710UG
Add to Cart
Add to Cart
Recombinant, human procathepsin V (amino acids 18-334) fused at the N-terminus to a CD33 signal sequence (amino acids 1-16) and at the C-terminus to a His·Tag™ sequence and expressed in NSO cells. Cat V is predominantly expressed in normal thymus, testis, and corneal epithelium. It is involved in the invariant chain degradation in thymic epithelial cells and its overexpression has been detected in myasthenia gravis. Abnormally wide spread expressions in mammary gland and colon have also been reported in breast and colonrectal carcinomas. Although it shows high amino acid identity with cathepsin L, these two cathepsins have very different tissue distribution and substrate specificity. As a result of glycosylation, this recombinant enzyme migrates ∽40kDa on SDS-PAGE under either reducing or non-reducing conditions. Requires activation prior to use.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 800mM NaCl, 25mM sodium acetate, 0.8M NaCl, pH 5.0.
Purification Method SDS-PAGE
Quantity 10 μg
Immunogen Cathepsin V
Storage Requirements To activate rhCathepsin V, pre-incubate it at 20μg/mL in 25mM NaOAc, 100mM NaCl, 5mM DTT, pH 5.5 at room temperature. Following initial thaw aliquot and freeze (−70°C).
Regulatory Status RUO
Cell Type NSO Cells
Conjugate His-Tag
Species Human
Recombinant Recombinant
Form Frozen
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.