Learn More
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Name | Human Recombinant Protein, CD40 Ligand/TRAP |
| Quantity | 10 μg |
| Source | E. coli |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
| Purity or Quality Grade | Greater than 98% by SDS-PAGE and HPLC analyses |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.