Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Centriprep™ Centrifugal Filter Units

Catalog No. 4302 Shop All MilliporeSigma Products
Change view
Click to view available options
MWCO:
3 kDa
Quantity:
24Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity
4302 3 kDa 24Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4302 Supplier MilliporeSigma™ Supplier No. 4302

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

For processing high-solute samples

Centriprep™ centrifugal filters are disposable ultrafiltration devices used for purifying, concentrating, and desalting biological samples in the 2–15 mL volume range. These complete, ready-to-use ultrafiltration devices are designed for operation in most centrifuges that can accommodate 50 mL centrifuge tubes. They are easy to use and offer a high flow rate. Centriprep™ devices consist of a sample container with a twist-lock cap, a filtrate collector containing a low-adsorptive, Ultracel™ regenerated cellulose membrane and an air-seal cap for sample isolation.

Features & Benefits:

  • For use with samples containing high levels of solutes
  • Unique inverse flow mode of operation with large deadstop
  • Fast sample processing
  • Available with Ultracel™ membranes: (3, 10, 30 and 50 kDa NMWL)
  • Fits standard swinging-bucket rotor for 50 mL tubes

Applications:Concentration and Purification of Particle-laden Solutions, Solutions with High Solute Concentration, Separation of Low MW Solutes from Fermentation Broths, Cell Culture Media and Lysates

TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Material (Membrane) Ultracel-YM Regenerated Cellulose
MWCO 3 kDa
Sterility Non-sterile
Filtration Area 2.84 cm2
Housing Material Regenerated Cellulose (RC)
Quantity 24Pack
Type Centrifugal Filters
Volume (Metric) 12 to 15 mL
Length (Metric) 135 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.