Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ceruloplasmin, Human Plasma, Protein

Catalog No. 2397991MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-979-91MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-979-91MG Supplier MilliporeSigma™ Supplier No. 2397991MG
Add to Cart
Add to Cart
Native ceruloplasmin from human plasma. Serum copper transport and iron-oxidizing protein. Plays an important role in antioxidant protection against organic and inorganic oxygen radicals generated by iron and ascorbate.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 100mM KCl, 50mM potassium phosphate buffer, 20mM ε-aminocaproic acid, 5mM EDTA, pH 6.8.
Preparation Method H2O (1mg/mL)
Purification Method SDS-PAGE
Quantity 1 mg
Immunogen Ceruloplasmin
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock Solutions are stable for up to 4 months at −20°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.