Learn More
Description
- Includes ready-to-use adipogenesis inducers.
- Oil Red O staining enables spectrophotometric analysis of lipid accumulation.
Specifications
Specifications
| Content And Storage | Dexamethasone Solution, IBMX Solution and Insulin Solution should be stored at -20°C. Oil Red O Solution, Wash Solution, and Dye Extraction Solution should be stored at room temperature |
| Description | Adipogenesis Assay Kit |
| Detection Method | Chromogenic |
| Includes | Dye Extraction, BMX, Insulin Solution, Dexamethasone, Oil Red O, Wash solution |
| Quantity | 1 Kit |
| Type | Adipogenesis Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.