Learn More
MilliporeSigma™ Chemicon™ ApopTag™ Fluorescein In Situ Apoptosis Detection Kit
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Content And Storage | Store the kit at -15 to -25°C until the first use. After the first use, if the kit will be used within three months, store the TdT Enzyme at -15 to -25°C and store remaining components at 2-8°C |
| Description | ApopTag fluorescein in situ apoptosis detection kit |
| Detection Method | Fluorescence |
| For Use With (Application) | Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Includes | Affinity purified sheep polyclonal antibody, equilibration buffer, reaction buffer, TdT enzyme, stop/wash buffer, Anti-Digoxigenin-Fluorescein, plastic coverslips |
| Quantity | 40 Assays |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.