Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Cell Comb™ Scratch Assay Kit

Catalog No. 1710191 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit (6 Assays)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-101-91 1 Kit (6 Assays)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-101-91 Supplier MilliporeSigma™ Supplier No. 1710191
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Western Blotting, Invasion Assay, Migration, Cell Based Assays, Activity Assay, Mass Spectrometry Sample Prep

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store Cell Combs and Cell Culture Plates at room temperature
Description Cell Comb Scratch Assay Kit
For Use With (Application) Western Blotting, Invasion Assay, Migration, Cell Based Assays, Activity Assay, Mass Spectrometry Sample Prep
Includes Cell Combs: Quantity of 6 individually packaged, disposable combs, Rectangular Cell Culture Plates: Quantity of 6 individually packaged, cell culture-treated 86mM x 128mM plates
Quantity 1 Kit (6 Assays)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.