Learn More
MilliporeSigma™ Chemicon™ CpG WIZ™ DAP-Kinase-Methylation Specific PCR Assay
Shop All MilliporeSigma ProductsDescription
- Methylation-specific PCR format.
Specifications
Specifications
| Description | CpG WIZ DAP-Kinase-Methylation specific PCR assay |
| For Use With (Application) | PCR |
| Includes | Universal 10X PCR buffer, U primer set 7.5μm, M primer set 7.5μm, W primer set 7.5μm, U control DNA, M control DNA, W control DNA |
| Quantity | 25 Assays |