Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ CpG WIZ™ SOCS1-Methylation Specific PCR Assay

Catalog No. S7809 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7809 25 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7809 Supplier MilliporeSigma™ Supplier No. S7809
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For PCR amplification after bisulfite modification of DNA samples

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -15°C to -25°C
Description CpG WIZ SOCS1-Methylation specific PCR assay
For Use With (Application) PCR
Includes Universal 10X PCR buffer, U primer set 5μm, M primer set 5μm, W primer set 5μm, U control DNA, M control DNA, W control DNA
Quantity 25 Assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.