Learn More
MilliporeSigma™ Chemicon™ CpG WIZ™ VHL-Methylation Specific PCR Assay
Shop All MilliporeSigma ProductsDescription
- Primer sets distinguish methylated and unmethylated DNA after bisulfite treatment.
- Supports mapping of methylation patterns across CpG regions.
Specifications
Specifications
| Description | CpG WIZ VHL-methylation specific PCR assay |
| For Use With (Application) | PCR |
| Quantity | 25 Assays |
| Type | DNA Assay Kit |