Learn More
MilliporeSigma™ Chemicon™ ECM Cell Adhesion Array Kit, Fluorimetric
Shop All MilliporeSigma ProductsDescription
- Microtiter plate array with homogeneous fluorescence detection.
- Enables quantitative comparison of multiple samples or cell lines.
Specifications
Specifications
| Description | ECM Cell Adhesion Array Kit |
| Detection Method | Fluorescence |
| For Use With (Application) | For assessing specific cell surface integrins, monitoring in vitro cell differentiation, or screening potential cell adhesion promoters/inhibitors |
| Quantity | 1/Pk. |
| Type | Cell-Based Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.