Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Fibrin Gel In vitro Angiogenesis Assay Kit

Catalog No. ECM630 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM630 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM630 Supplier MilliporeSigma™ Supplier No. ECM630
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Activity Assay

  • Fibrin gel supports evaluation of endothelial tube formation.
  • Suitable for studying multiple stages of angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store unopened kit materials at -20°C or -70°C
Description Fibrin gel in vitro angiogenesis assay kit
For Use With (Application) Activity Assay
Includes 100X positive control angiogenic supplement, fibrinogen, thrombin solution
Quantity 1 Kit
Type Cell-Based Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.