Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon Human Interferon-gamma Recombinant Protein

Catalog No. IF002 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IF002 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IF002 Supplier MilliporeSigma™ Supplier No. IF002
Add to Cart
Add to Cart

Specifications

For Use With (Application) Cell Culture
Name Human Interferon-g Recombinant Protein
Quantity 100 μg
Source E. coli
Recombinant Recombinant
Purity or Quality Grade Greater than 98% by SDS-PAGE and HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.