Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Human TWEAK

Catalog No. GF102 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF102 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF102 Supplier MilliporeSigma™ Supplier No. GF102
Add to Cart
Add to Cart

Secreted ligand in tumor necrosis factor family that weakly induces apoptosis

  • Induces interleukin-8 synthesis in many cell lines
  • Source: E.Coli
  • Species: Human
  • Purity: >98% by SDS-PAGE and HPLC analysis
  • Sterile filtered
  • Lyophilized from 10mM NaP, pH: 7.5 and 0.1M L-Arg

Recommended Storage

Store at -20° C

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Quantity 25 μg
Source E.coli
Storage Requirements Store at -20°C
Gene Symbol MGC20669, TNFSF12, MGC129581, DR3LG, TWEAK, APO3L
Species Human
Sterility Sterile
Protein Family Tumor necrosis factor family
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.