Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ In vitro Osteogenesis Assay Kit

Catalog No. ECM810 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM810 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM810 Supplier MilliporeSigma™ Supplier No. ECM810
Add to Cart
Add to Cart

Efficient in vitro differentiation, end-point determination, quantification of MC3T3-E1 cells to mature osteoblasts

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Kit components require two different storage temperatures: 2-8°C, -20°C
Description In vitro Osteogenesis assay kit
Detection Method Chromogenic
For Use With (Application) Efficient in vitro differentiation, end-point determination, quantification of MC3T3-E1 cells to mature osteoblasts
Includes 10X ARS dilution buffer, ascorbic acid 2-phosphate solution (500X), glycerol 2- phosphate solution (100X), melatonin Solution, Alizarin Red S stain solution (1X), 10% acetic acid, 10% ammonium hydroxide
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.