Learn More
MilliporeSigma™ Chemicon™ In Vitro Vascular Permeability Imaging Assay (Green)
Shop All MilliporeSigma ProductsDescription
- Supports compound studies of endothelial monolayer integrity.
- Includes reagents for fluorescence imaging.
Specifications
Specifications
| Description | In Vitro Vascular Permeability Imaging Assay (Green) kit |
| Detection Method | Fluorescence |
| Includes | Donkey anti-Mouse IgG, Cy3 Conjugate Secondary, Poly-L-Lysine, 2X, DAPI, Glutaraldehyde, 16X, Biotin-Gelatin, Fluorescein-Streptavidin, VE Cadherin Primary Antibody |
| Quantity | 1 Kit (32 Assays) |