Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ LentiBrite™ GFP-Vimentin Lentiviral Biosensor

Catalog No. 1710152 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Vial (minimum of 3 x 108 IFU/mL)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-101-52 1 Vial (minimum of 3 x 108 IFU/mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-101-52 Supplier MilliporeSigma™ Supplier No. 1710152
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Track single cell and cell population migration (migration assay, cell culture), Transfection, Immunofluorescence, Immunocytochemistry

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Lentivirus is stable for at least 4 months from date of receipt when stored at -80°C.
Description GFP-Vimentin Lentiviral Biosensor
Detection Method Fluorescence
For Use With (Application) Track single cell and cell population migration (migration assay, cell culture), Transfection, Immunofluorescence, Immunocytochemistry
Includes TagGFP2-Vimentin Lentivirus: One vial containing 25μL of lentiviral particles at a minimum of 3 x 10E8 infectious units (IFU) per mL; Promoter: EF-1 (Elongation Factor-1)
Quantity 1 Vial (minimum of 3 x 108 IFU/mL)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.