Learn More
MilliporeSigma™ Chemicon™ LentiBrite™ RFP-LC3 Control Mutant Lentiviral Biosensor
Shop All MilliporeSigma ProductsDescription
- Fluorescent lentiviral particles serve as a negative control for live-cell autophagy analysis.
Specifications
Specifications
| Content And Storage | Lentivirus is stable for at least 4 months from date of receipt when stored at -80°C. |
| Description | RFP-LC3 Control Mutant Lentiviral Biosensor |
| Detection Method | Fluorescence |
| For Use With (Application) | Immunocytochemistry, Transfection, Immunofluorescence |
| Includes | TagRFP-LC3-G120A Lentivirus: One vial containing 25μL of lentiviral particles at a minimum of 3 x 10E8 infectious units (IFU) per mL; Promoter: EF-1 (Elongation Factor-1) |
| Quantity | 1 Vial (minimum of 3 x 108 IFU/mL) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.