Learn More
MilliporeSigma™ Chemicon™ LentiBrite™ RFP-Vimentin Lentiviral Biosensor
Shop All MilliporeSigma ProductsDescription
- Prepackaged fluorescent lentiviral biosensor.
- Supports visualization in living cells.
Specifications
Specifications
| Content And Storage | Lentivirus is stable for at least 4 months from date of receipt when stored at -80°C. |
| Description | RFP-Vimentin Lentiviral Biosensor |
| Detection Method | Fluorescence |
| For Use With (Application) | Track single cell and cell population migration (migration assay, cell culture), Transfection, Immunofluorescence, Immunocytochemistry |
| Includes | TagRFP-Vimentin Lentivirus: One vial containing 25μL of lentiviral particles at a minimum of 3 x 10E8 infectious units (IFU) per mL; Promoter: EF-1 (Elongation Factor-1) |
| Quantity | 1 Vial (minimum of 3 x 108 IFU/mL) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.