Learn More
Description
- Stimulates growth of activated CD4+ and CD8+ T cells and NK cells
- Disulfide linked heterodimeric protein
- Molecular weight: 75kd
- Sterile filtered
- Lyophilized from 0.5x PBS, pH: 7.2
- Purity: >95% by SDS-PAGE
- Source: CHO cells (Chinese Hamster Ovarian cells)
- Species: Mouse
Recommended Storage
Store at -20°C
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Molecular Weight (g/mol) | 75kd |
| Quantity | 10 μg |
| Storage Requirements | Store at -20°C |
| Product Type | Mouse Interleukin-12 Recombinant Protein |
| Species | Mouse |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95% by SDS-PAGE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.