Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Neurite Outgrowth Assay Kits

Catalog No. NS225 Shop All MilliporeSigma Products
Change view
Click to view available options
Description:
Neurite Outgrowth Assay Kit (1μm)
Neurite Outgrowth Assay Kits (3μm)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
NS225 Neurite Outgrowth Assay Kit (1μm)
NS220 Neurite Outgrowth Assay Kits (3μm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NS225 Supplier MilliporeSigma™ Supplier No. NS225
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uses Millicell cell culture inserts (chambers) containing permeable membrane

  • Separates projecting neurites from cell bodies across a microporous membrane.
  • Enables analysis of the neurite population on the membrane underside.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Neurite Outgrowth Assay Kit (1μm)
Detection Method Chromogenic
Includes Forceps (1 Each), Neurite Outgrowth Plate Assembly, Neurite Stain Solution (one 20mL bottle), Neurite Stain Extraction Buffer (one 20mL bottle), Neurite Outgrowth Assay Plate (two 24-well plates), 50 Cotton swabs
Quantity 12 Assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.