Learn More
Description
- Includes Alizarin Red staining and extraction reagents.
- Supports quantitative analysis with a standard plate reader.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Osteogenesis quantitation kit |
| Detection Method | Chromogenic |
| For Use With (Application) | Studying effects of growth factors, drugs and toxic agents on bone formation, screening candidate osteogenic pharmaceuticals |
| Includes | 10X ARS dilution buffer, Alizarin Red S stain solution (1X), 10% acetic acid, 10% ammonium hydroxide |
| Quantity | 1 Kit |
| Type | Quantitation Kit |
| Druggable Target | Cell-based Assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.