Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ OxyIHC™ Oxidative Stress Detection Kit

Catalog No. S7450 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7450 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7450 Supplier MilliporeSigma™ Supplier No. S7450
Add to Cart
Add to Cart

Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description OxyIHC Oxidative Stress Detection Kit
Detection Method Chromogenic
For Use With (Application) Immunohistochemistry
Includes Derivatization control solution, 10X wash buffer, 10X antigen retrieval buffer, 5X blocking buffer, primary antibody, DAB-A, DAB-B, antibody diluent, biotinylated secondary antibody, streptavidin conjugated HRP, hematoxylin, DNPH solution
Quantity 50 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.