Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Pseudopodia Purification Kit

Catalog No. ECM660 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM660 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM660 Supplier MilliporeSigma™ Supplier No. ECM660
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isolate and quantitate extending or retracting pseudopodia from cell body

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store kit and materials at 2-8°C until their expiration date
Description Pseudopodia Purification Kit
For Use With (Application) Isolate and quantitate extending or retracting pseudopodia from cell body
Includes Pseudopodia purification inserts, western blot lysis/Immunoprecipitation buffer, cell scrapers, forceps
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.