Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ QCM Chemotaxis Cell Migration Assay, 24-well (5μm), Chromogenic

Catalog No. ECM506 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM506 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM506 Supplier MilliporeSigma™ Supplier No. ECM506
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Activity Assay

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description QCM Chemotaxis Cell Migration Assay
Detection Method Chromogenic
For Use With (Application) Activity Assay
Includes Forceps, Sterile 5μm 24-well Cell migration plate, two 24-well plates w/12 inserts/plate, Cell Stain, Extraction buffer, Cotton Swaps
Quantity 1/Pk.
Type Cell-Based Assay Kit
Druggable Target Cell-based Assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.