Learn More
MilliporeSigma™ Chemicon™ QCM Laminin Migration Assay, 24-well, Colorimetric
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Description | Laminin Migration Assay |
| Detection Method | Colorimetric |
| For Use With (Application) | For examine cell migration dependent upon the presence of laminin |
| Includes | Laminin, 24-well cell migration plate assembly, BSA 30% solution, Cell Stain Solution, Extraction Buffer, 24-well stain receiver plate, 96-Well Stain Quantitation Plate, Swabs, Forceps |
| Quantity | 1/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.